Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP4S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114334
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114334 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114334 Supplier Invitrogen™ Supplier No. PA5114334
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody specifically recognizes AP4S1 in Arabidopsis thaliana.

This gene encodes a member of the adaptor complexes small subunit protein family. These proteins are components of the heterotetrameric adaptor protein complexes, which play important roles in the secretory and endocytic pathways by mediating vesicle formation and sorting of integral membrane proteins. The encoded protein is the small subunit of adaptor protein complex-4, which is associated with both clathrin- and nonclathrin-coated vesicles. Mutations in this gene are associated with spastic quadriplegic cerebral palsy-6. Alternatively spliced transcript variants encoding multiple isoforms have been observed for this gene, and a pseudogene of this gene is located on the long arm of chromosome 6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP4S1
Applications ELISA, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene Alias adapter-related protein complex 4 subunit sigma-1; adaptor related protein complex 4 sigma 1 subunit; adaptor related protein complex 4 subunit sigma 1; adaptor related protein complex 4, sigma 1 subunit; adaptor-related protein complex 4 subunit sigma-1; adaptor-related protein complex 4, sigma 1 subunit; adaptor-related protein complex AP-4, sigma 1; AI314282; AP4; AP-4 adapter complex subunit sigma-1; AP-4 adaptor complex subunit sigma-1; AP-4 complex subunit sigma-1; AP47B; Ap4s1; ap4s1 protein; CLA20; CLAPS4; clathrin-associated/assembly/adaptor protein, sigma 4; CPSQ6; LOC618267 protein; si:dz115o7.1; si:dz234g15.5; sigma-1 subunit of AP-4; Sigma-4-adaptin; sigma4-adaptin; SPG52; zgc:136677
Host Species Rabbit
Immunogen Synthetic peptide within thaliana AP-4 complex subunit sigma aa 51-100.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Plant
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.