Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ AP5M1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA563174
Description
Immunogen sequence: PLQDILVHPC VTSLDSAILT SSSIDAMDDS AFSGPYKFPF TPPLESFNLC FYTSQVPVPP ILGFYQMKEE EVQLRITI Highest antigen sequence identity to the following orthologs: Mouse - 87%, Rat - 86%.
As part of AP-5, a probable fifth adaptor protein complex it may be involved in endosomal transport. According to PubMed:18395520, it may play a role in cell death. [UniProt].
Specifications
AP5M1 | |
Polyclonal | |
Unconjugated | |
AP5M1 | |
adapter-related protein complex 5 mu subunit; adapter-related protein complex 5 subunit mu-1; adaptor related protein complex 5 mu 1 subunit; adaptor related protein complex 5, mu 1 subunit; adaptor-related protein complex 5 subunit mu-1; adaptor-related protein complex 5, mu 1 subunit; AP-5 complex subunit mu; AP-5 complex subunit mu-1; AP5M1; C14orf108; MHD domain-containing death-inducing protein; Mu-2 related death-inducing; MU-2/AP1M2 domain containing, death-inducing; mu-2-related death-inducing protein; Mu5; MuD; MUDENG; Putative HIV-1 infection-related protein | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
55745 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9H0R1 | |
AP5M1 | |
Recombinant protein corresponding to Human AP5M1. Recombinant protein control fragment (Product #RP-101780). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction