Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP5M1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563401
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563401 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563401 Supplier Invitrogen™ Supplier No. PA563401
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: CPFGTLLDAN LQNSLDNTNF ASVTQPQKQP AWKTGTYKGK PQVSISITEK VKSMQYDKQG IADTWQVVGT VTCKCDLEGI MPNVTISLSL PT Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 85%.

As part of AP-5, a probable fifth adaptor protein complex it may be involved in endosomal transport. According to PubMed:18395520, it may play a role in cell death. [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP5M1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP5M1
Gene Accession No. Q9H0R1
Gene Alias adapter-related protein complex 5 mu subunit; adapter-related protein complex 5 subunit mu-1; adaptor related protein complex 5 mu 1 subunit; adaptor related protein complex 5, mu 1 subunit; adaptor-related protein complex 5 subunit mu-1; adaptor-related protein complex 5, mu 1 subunit; AP-5 complex subunit mu; AP-5 complex subunit mu-1; AP5M1; C14orf108; MHD domain-containing death-inducing protein; Mu-2 related death-inducing; MU-2/AP1M2 domain containing, death-inducing; mu-2-related death-inducing protein; Mu5; MuD; MUDENG; Putative HIV-1 infection-related protein
Gene Symbols AP5M1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP5M1. Recombinant protein control fragment (Product #RP-101779).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55745
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.