Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ AP5S1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583663
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583663 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583663 Supplier Invitrogen™ Supplier No. PA583663
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ENLLLAEGTL RLLTRLLLDH LRLLAPSTSL LLRADRIEGI LTRFLPHGQL LFLNDQFVQG LEKEFSAAW Highest antigen sequence indentity to the following orthologs: Mouse - 90%, Rat - 90%.

AP5S1 is a protein coding gene. As part of AP-5, a probable fifth adaptor protein complex it may be involved in endosomal transport. According to PubMed:20613862, it is required for efficient homologous recombination DNA double-strand break repair.
TRUSTED_SUSTAINABILITY

Specifications

Antigen AP5S1
Applications Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene AP5S1
Gene Accession No. Q9NUS5
Gene Alias adapter-related protein complex 5 sigma subunit; adaptor related protein complex 5 sigma 1 subunit; adaptor related protein complex 5, sigma 1 subunit; adaptor-related protein complex 5 sigma subunit; adaptor-related protein complex 5, sigma 1 subunit; AP-5 complex subunit sigma-1; AP5S1; C20orf29; FLJ11168; protein C20orf29; sigma5
Gene Symbols AP5S1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human AP5S1. Recombinant protein control fragment (Product #RP-108361).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 55317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.