Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APAAP Complex Monoclonal Antibody (WR31)
GREENER_CHOICE

Catalog No. PIMA180405
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA180405 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA180405 Supplier Invitrogen™ Supplier No. MA180405
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

MA1-80405 detects APAAP Complex: an alkaline phosphatase linked to a monoclonal anti-alkaline phosphatase antibody. APAAP complexes are designed for use as detection reagents in immunohistochemistry procedures. They are used in combination with a suitable bridging antibody and appropriate visualization system.

APAAP complexes are designed for use as detection reagents in immunohistochemistry procedures. They are used in combination with a suitable bridging antibody and appropriate visualization system. APAAP is a complex of anti-alkaline phosphatase antibodies with alkaline phosphatase.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APAAP Complex
Applications Immunohistochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone WR31
Conjugate Unconjugated
Formulation cell culture media with 0.09% sodium azide
Gene Alias Alkaline Phosphatase Complex
Host Species Mouse
Immunogen Bovine alkaline phosphatase.
Purification Method purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Chemical
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1
Target Molecule Chemical
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.