Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APAF-1 Antibody (13F11), Alexa Fluor™ 532, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280093AF532
This item is not returnable.
View return policy
Description
APAF-1 Monoclonal antibody specifically detects APAF-1 in Mouse, Rat samples. It is validated for Western Blot, ELISA, ImmunofluorescenceSpecifications
APAF-1 | |
Monoclonal | |
Alexa Fluor 532 | |
APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
Recombinant mouse APAF-1 (aa 1-97) containing the N-terminal CARD domain. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
317 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunofluorescence | |
13F11 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction