Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APAF-1 Antibody (2E12), DyLight 550, Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP280092R
This item is not returnable.
View return policy
Description
APAF-1 Monoclonal antibody specifically detects APAF-1 in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
APAF-1 | |
Monoclonal | |
DyLight 550 | |
APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
Recombinant human APAF-1 (aa 1-464) containing the N-terminal CARD and CED-4 homologous domains. | |
0.1 mL | |
Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
317 | |
Store at 4°C in the dark. | |
IgG2a κ |
Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
2000000000000 | |
50mM Sodium Borate | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction