Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APAF-1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody has been used in 2 publications
Supplier: Novus Biologicals NBP177000
Description
APAF-1 Polyclonal specifically detects APAF-1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
APAF-1 | |
Polyclonal | |
Unconjugated | |
PBS with 0.02% Sodium Azide | |
APAF-1apoptotic protease activating factor 1, apoptotic peptidase activating factor, apoptotic peptidase activating factor 1, apoptotic protease activating factor, apoptotic protease-activating factor 1, CED4, DKFZp781B1145, KIAA0413 | |
Rabbit | |
130 kDa | |
0.1 mg | |
Apoptosis, Cancer, Cell Biology, Mitochondrial Markers, Tumor Suppressors | |
317 | |
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
1 mg/ml | |
Western Blot 1 μg/mL, ELISA 1:100-1:2000, Immunohistochemistry 1 μg/mL, Immunocytochemistry/Immunofluorescence 10 μg/mL, Immunoprecipitation, Immunohistochemistry-Paraffin 1 μg/mL | |
AAC51678 | |
APAF1 | |
Antibody was raised against a peptide corresponding to amino acids near the amino terminus of human Apaf1. The sequences of the immunogenic peptide are identical between human and mouse. The immunogen is located within the first 50 amino acids of Apaf1. Amino Acid Squence: HREALEKDIKTSYIMDH | |
Affinity Purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction