Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APAF1 Monoclonal Antibody (2H9A1)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548371
Description
APAF1 Monoclonal Antibody for Western Blot, ICC/IF, Flow, ELISA
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
Specifications
APAF1 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
O14727 | |
APAF1 | |
Purified recombinant fragment of human APAF1 (AA: 1138-1237) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2b |
ELISA, Flow Cytometry, Western Blot, Immunocytochemistry | |
2H9A1 | |
Unconjugated | |
APAF1 | |
6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413 | |
Mouse | |
Protein G | |
RUO | |
317 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction