Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APAF1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583190
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583190 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583190 Supplier Invitrogen™ Supplier No. PA583190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PLVVSLIGAL LRDFPNRWEY YLKQLQNKQF KRIRKSSSYD YEALDEAMSI SVEMLREDIK DYYTDLSILQ KDVKVPTKVL CILWDMETEE VEDILQEFVN KSLLFCDR Highest antigen sequence indentity to the following orthologs: Mouse - 95%, Rat - 94%.

APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APAF1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APAF1
Gene Accession No. O14727
Gene Alias 6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413
Gene Symbols APAF1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APAF1. Recombinant protein control fragment (Product #RP-95285).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 317
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.