Learn More
Invitrogen™ APAF1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5120054
Description
Positive test controls include: Jurkat, 293T, Mouse testis. The target is usually found in the following locations: Cytoplasm. Immunogen sequence: PHTLSPDQED CMYWYNFLAY HMASAKMHKE LCALMFSLDW IKAKTELVGP AHLIHEFVEY RHILDEKDCA VSENFQEFLS LNGHLLGRQP FPNIVQLGLC EPETSEVYQQ AKLQAKQEVD NGMLYLEWIN K.
APAF1 gene encodes a cytoplasmic protein that initiates apoptosis. This protein contains several copies of the WD-40 domain, a caspase recruitment domain (CARD), and an ATPase domain (NB-ARC). Upon binding cytochrome c and dATP, this protein forms an oligomeric apoptosome. The apoptosome binds and cleaves caspase 9 preproprotein, releasing its mature, activated form. Activated caspase 9 stimulates the subsequent caspase cascade that commits the cell to apoptosis. Alternative splicing results in several transcript variants encoding different isoforms.
Specifications
APAF1 | |
Polyclonal | |
Unconjugated | |
APAF1 | |
6230400I06Rik; APAF1; APAF-1; Apaf1l; apoptotic peptidase activating factor 1; apoptotic protease activating factor 1; apoptotic protease-activating factor 1; CED4; DKFZp781B1145; fog; forebrain overgrowth; KIAA0413; mKIAA0413 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
11783, 317, 78963 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Western Blot, Immunocytochemistry | |
1.22 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
O14727, O88879, Q9EPV5 | |
APAF1 | |
Recombinant protein (or fragment). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
Your input is important to us. Please complete this form to provide feedback related to the content on this product.