Learn More
Eisco™ Apatite Mineral Specimen
SCI_ED
Catalog No. S26675 Shop All Eisco Products
Change view
Click to view available options
Quantity:
10/Pk.
Catalog No. Quantity
S26675 10/Pk.
1 options
Catalog No. S26675 Supplier Eisco™ Supplier No. ESNG0065PK10
Add to Cart
Add to Cart
Apatite Mineral Specimen ideal for use in lessons about the Mohs hardness scale.
  • Used as a hardness standard of 5 (Mohs scale)
  • Good example of mineral fracture
  • Correlates well to Next Generation Science Standards (NGSS) Evidence Statements
Specifications
Product Type Metamorphic Rock
Quantity 10/Pk.
Specimen Size 2 to 3 cm
Weight (Metric) 0.2 kg
Specimen Type Apatite