Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA1 Antibody, Novus Biologicals™

Goat Polyclonal Antibody
Supplier: Novus Biologicals NBP106015
Description
APBA1 Polyclonal specifically detects APBA1 in Human samples. It is validated for Western Blot, Peptide ELISA.Specifications
APBA1 | |
Polyclonal | |
Unconjugated | |
Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide | |
Adapter protein X11alpha, amyloid beta (A4) precursor protein-binding, family A, member 1, amyloid beta A4 precursor protein-binding family A member 1, D9S411Eadaptor protein X11alpha, LIN10, Mint-1, MINT1amyloid beta (A4) precursor protein-binding, family A, member 1 (X11), Neuronal Munc18-1-interacting protein 1, Neuron-specific X11 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, UROP11, X11A, X11X11ALPHA | |
Goat | |
Affinity Purified | |
RUO | |
Primary | |
Human. Predicted cross-reactivity based on sequence identity: Mouse, Rat. | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, ELISA | |
0.5 mg/mL | |
Western Blot 0.1 - 0.3 ug/ml, Peptide ELISA Detection limit 1:16000 | |
NP_001154.2 | |
APBA1 | |
Peptide with sequence C-EASHPSQDGKRQYK corresponding to internal region according to NP_001154.2. | |
0.1 mg | |
Membrane Trafficking and Chaperones | |
320 | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction