Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA1 Polyclonal Antibody

APBA1 Polyclonal Antibody
Supplier: Thermo Scientific PA518702
Description
APBA1 Polyclonal Antibody for Western Blot

Specifications
APBA1 | |
Polyclonal | |
Unconjugated | |
APBA1 | |
6430513E09Rik; adapter protein X11alpha; adaptor protein X11alpha; amyloid beta (A4) precursor protein binding, family A, member 1; amyloid beta (A4) precursor protein-binding family A APBA1: amyloid beta (A4) precursor protein-binding family A member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, APBA1: amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, member 1; amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta A4 precursor protein-binding family A member 1; amyloid beta precursor protein binding family A member 1; Amyloid-beta A4 precursor protein-binding family A member 1; APBA1; D9S411E; LIN10; Lin-10; Mint; MINT1; Mint-1; neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; Urop11; UROP11-110; X11; X11A; X11ALPHA | |
Goat | |
Ammonium sulfate precipitation | |
RUO | |
320 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
Q02410 | |
APBA1 | |
Synthetic peptide sequence (EASHPSQDGKRQYK) corresponding to the internal amino acids of APBA1 (aa 573-587). | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction