Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBA1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA534418
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA534418 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA534418 Supplier Thermo Scientific Supplier No. PA534418
Add to Cart
Add to Cart

APBA1 Polyclonal Antibody

APBA1 Polyclonal Antibody for Western Blot, IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen APBA1
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene APBA1
Gene Accession No. B2RUJ5, O35430, Q02410
Gene Alias 6430513E09Rik; adapter protein X11alpha; adaptor protein X11alpha; amyloid beta (A4) precursor protein binding, family A, member 1; amyloid beta (A4) precursor protein-binding family A APBA1: amyloid beta (A4) precursor protein-binding family A member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, APBA1: amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta (A4) precursor protein-binding, family A, member 1; amyloid beta (A4) precursor protein-binding, family A, member 1 (X11); amyloid beta A4 precursor protein-binding family A member 1; amyloid beta precursor protein binding family A member 1; Amyloid-beta A4 precursor protein-binding family A member 1; APBA1; D9S411E; LIN10; Lin-10; Mint; MINT1; Mint-1; neuronal Munc18-1-interacting protein 1; Neuron-specific X11 protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; Urop11; UROP11-110; X11; X11A; X11ALPHA
Gene Symbols APBA1
Host Species Rabbit
Immunogen A 19 amino acid synthetic peptide near the amino terminus of human APBA1.
Purification Method Antigen Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 319924, 320, 83589
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.