Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APBA2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA547830
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA547830 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA547830 Supplier Invitrogen™ Supplier No. PA547830
Add to Cart
Add to Cart

Sheep Polyclonal Antibody

This antibody detects human APBA2 in direct ELISAs and human, mouse and rat in Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) APBA1 and rhAPBA3 is observed. Reconstitute at 0.2 mg/mL in sterile PBS.

APBA2, a member of the X11 protein family, is a phosphotyrosine-binding domain protein and is a neuronal adapter protein that interacts with amyloid precursor protein (APP) and neuritic plaques in the brains of patients with Alzheimer's disease. It stabilizes APP and inhibits production of proteolytic APP fragments including the Abeta peptide that is deposited in the brains of Alzheimer's disease patients. APBA2 is believed to be involved in signal transduction processes and is also regarded as a putative vesicular trafficking protein in the brain that can form a complex with the potential to couple synaptic vesicle exocytosis to neuronal cell adhesion. Recent reports suggest that it may also be a candidate gene for autism.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APBA2
Applications Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and No Preservative
Gene APBA2
Gene Accession No. O35431, P98084, Q99767
Gene Alias Adapter protein X11beta; amyloid beta (A4) precursor protein-binding family A member 2 (X11-like); amyloid beta (A4) precursor protein-binding, family A, member 2; amyloid beta (A4) precursor protein-binding, family A, member 2 (X11-like); amyloid beta A4 precursor protein-binding family A member 2; amyloid beta precursor protein binding family A member 2; Amyloid-beta A4 precursor protein-binding family A member 2; Apba2; D15S1518E; HsT16821; LIN-10; MGC:14091; Mint 2; Mint2; mint-2; mXllL; Neuronal Munc18-1-interacting protein 2; neuron-specific X11L protein; phosphotyrosine-binding/-interacting domain (PTB)-bearing protein; X11-BETA; X11L; X11-like; X11-like protein; XllL
Gene Symbols APBA2
Host Species Sheep
Immunogen E. coli-derived recombinant human APBA2 Ala2-Leu165.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11784, 321, 83610
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.