Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APBA3/Mint-3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB1203450
Description
APBA3/Mint-3 Polyclonal specifically detects APBA3/Mint-3 in Mouse, Rat samples. It is validated for Western Blot.Specifications
APBA3/Mint-3 | |
Polyclonal | |
Unconjugated | |
PBS with 0.05% Sodium Azide | |
APBA3 | |
Synthetic peptide corresponding to residues M(1) E F L P E P Q H P P G P P T M D L E(19) of rat Mint3. | |
100 μg | |
Membrane Trafficking and Chaperones, Vision | |
9546 | |
Mouse, Rat | |
IgG |
Western Blot | |
1 mg/mL | |
Western Blot 2 ug/ml | |
Adapter protein X11gamma, amyloid beta (A4) precursor protein-binding, family A, member 3, amyloid beta (A4) precursor protein-binding, family A, member 3 (X11-like 2), MGC:15815, mint3, Mint-3, Neuronal Munc18-1-interacting protein 3, Neuron-specific X11L2 protein, phosphotyrosine-binding/-interacting domain (PTB)-bearing protein, X11L2amyloid beta A4 precursor protein-binding family A member 3, X11-like 2 protein | |
Rabbit | |
Affinity Purified | |
RUO | |
Primary | |
NB120-3450 detects munc-18 interacting protein 3 (Mint3) from mouse cells and rat brain samples. | |
Store at -20C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction