Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB1IP Monoclonal Antibody (OTI6E3), TrueMAB™, OriGene
SDP

Catalog No. 501682653
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2653 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2653 Supplier OriGene Supplier No. TA809230
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RIAM appears to function in the signal transduction from Ras activation to actin cytoskeletal remodeling. Suppresses insulin-induced promoter activities through AP1 and SRE. Mediates Rap1-induced adhesion.

Specifications

Antigen APBB1IP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone OTI6E3
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene APBB1IP
Gene Accession No. Q7Z5R6
Gene Alias INAG1, PREL1, RARP1, RIAM
Gene Symbols APBB1IP
Host Species Mouse
Immunogen Full length human recombit protein of human APBB1IP produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 54518
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.