Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB1IP Mouse anti-Human, Clone:7E7A3, Abnova™

Catalog No. 89333133 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-133 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-133 Supplier Abnova Corporation Supplier No. MAB16641
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against recombinant human APBB1IP.

Specifications

Antigen APBB1IP
Applications ELISA, Flow Cytometry, Western Blot
Classification Monoclonal
Clone 7E7A3
Conjugate Unconjugated
Description amyloid beta (A4) precursor protein-binding, family B, member 1 interacting protein
Dilution ELISA (1:10000)
Western Blot (1:500-1:2000)
Flow Cytometry (1:200-1:400)
The optimal working dilution should be determined by the end user.
Formulation In PBS (0.05% sodium azide)
Gene APBB1IP
Gene Alias INAG1, PREL1, RARP1, RIAM
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acid 1-151 of human APBB1IP from E. coli.
Quantity 100 μg
Gene ID (Entrez) 54518
Target Species Human
Content And Storage Store at 4°C. For long term storage store at -20°C.
Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.