Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APBB2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5101421
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5101421 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5101421 Supplier Invitrogen™ Supplier No. PA5101421
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total APBB2.

The protein encoded by this gene interacts with the cytoplasmic domains of amyloid beta precursor protein and amyloid beta precursor-like protein 2. This protein contains two phosphotyrosine binding domains, which are thought to function in signal transduction. Polymorphisms in this gene have been associated with Alzheimer's disease. Alternative splicing results in multiple transcript variants.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APBB2
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Apbb2
Gene Accession No. Q92870, Q9DBR4
Gene Alias 2310007D03Rik; amyloid beta (A4) precursor protein-binding, family B, member 2; amyloid beta (A4) precursor protein-binding, family B, member 2 (Fe65-like); amyloid beta A4 precursor protein-binding family B member 2; amyloid beta A4 precursor protein-binding, family B, member 2; amyloid beta precursor protein binding family B member 2; amyloid beta precursor protein-binding family B member 2; amyloid-beta A4 precursor protein-binding family B member 2; Apbb2; FE65L; FE65L1; Fe65-like 1; Protein Fe65-like 1; RGD1562438; Rirl1; TR2L; Zfra; zinc finger-like protein
Gene Symbols Apbb2
Host Species Rabbit
Immunogen A synthesized peptide derived from human APBB2(Accession Q92870), corresponding to amino acid residues Q493-E543.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11787, 305338, 323
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.