Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APBB2 Polyclonal Antibody, Bethyl Laboratories

Catalog No. 501566088
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-6088 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-6088 Supplier Bethyl Laboratories, Inc Supplier No. A303798AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

The protein encoded by this gene interacts with the cytoplasmic domains of amyloid beta precursor protein and amyloid beta precursor-like protein 2. This protein contains two phosphotyrosine binding domains, which are thought to function in signal transduction. Polymorphisms in this gene have been associated with Alzheimer′s disease. Alternative splicing results in multiple transcript variants.

Specifications

Antigen APBB2
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene APBB2
Gene Accession No. Q92870
Gene Alias FE65L, FE65L1, Fe65-like 1, amyloid beta A4 precursor protein-binding family B member 2, protein Fe65-like 1
Gene Symbols APBB2
Host Species Rabbit
Immunogen Between 708 and 758
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 323
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.