Learn More
APBB3 Mouse anti-Human, Clone: OTI5G3, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF503752

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The protein encoded by this gene is a member of the APBB protein family. It is found in the cytoplasm and binds to the intracellular domain of the Alzheimer′s disease beta-amyloid precursor protein (APP) as well as to other APP-like proteins. It is thought that the protein encoded by this gene may modulate the internalization of APP. Multiple transcript variants encoding several different isoforms have been found for this gene.Specifications
APBB3 | |
Monoclonal | |
Unconjugated | |
APBB3 | |
FE65L2, SRA | |
Mouse | |
Affinity Chromatography | |
RUO | |
10307 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
OTI5G3 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O95704 | |
APBB3 | |
Full length human recombit protein of human APBB3 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.