Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APBB3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA552167
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA552167 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA552167 Supplier Invitrogen™ Supplier No. PA552167
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AIILVNCDDD LWGDHSLEVE AGLPPGWRKI HDAAGTYYWH VPSGSTQWQR PTWELGDAED PGTGTEGIWG LRPPKGRSFS SLESSLDRSN SLSWYGGESY IQSMEPGAKC FAVRSL Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

The protein encoded by this gene is a member of the APBB protein family. It is found in the cytoplasm and binds to the intracellular domain of the Alzheimer's disease beta-amyloid precursor protein (APP) as well as to other APP-like proteins. It is thought that the protein encoded by this gene may modulate the internalization of APP. Multiple transcript variants encoding several different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APBB3
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene APBB3
Gene Accession No. O95704
Gene Alias amyloid beta (A4) precursor protein-binding, family B, member 3; amyloid beta A4 precursor protein-binding family B member 3; amyloid beta precursor protein binding family B member 3; amyloid precursor interacting protein; Amyloid-beta A4 precursor protein-binding family B member 3; APBB3; Fe65L2; fe65-like protein 2; Protein Fe65-like 2; Rirl2; SRA; TR2S
Gene Symbols APBB3
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APBB3. Recombinant protein control fragment (Product #RP-88822).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10307
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.