Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC Antibody (KT44), Novus Biologicals™

Rat Monoclonal Antibody
Supplier: Novus Biologicals NBP323227
Description
APC Monoclonal antibody specifically detects APC in Human, Mouse, Rat samples. It is validated for Western Blot, ImmunofluorescenceSpecifications
APC | |
Monoclonal | |
Unconjugated | |
PBS | |
Rat | |
Protein G purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, Immunofluorescence | |
KT44 | |
Western Blot 1:250, Immunocytochemistry/Immunofluorescence 1:50 | |
adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
Clone KT44 was generated from a recombinant protein containing amino acid residues from the central region mouse APC. This sequence is highly conserved in human and rat APC, and has low homology to APC2. [UniProt# Q61315] | |
0.1 mL | |
Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
324 | |
Store at 4°C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction