Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC Antibody (OTI2B8) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP270183
Description
APC Monoclonal specifically detects APC in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
APC | |
Monoclonal | |
LYOPH | |
Western Blot 1:2000, Immunohistochemistry 1:150 | |
adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC | |
Mouse | |
311.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI2B8 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
APC | |
Human recombinant protein fragment corresponding to amino acids 955-1382 of human APC (NP_001120983) produced in E.coli. | |
Affinity Purified | |
Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway | |
324 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction