Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC-Cy7 Mouse Anti-Human CD29 (Integrin β1), BD Pharmingen™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 569941
Description
CD29 (Integrin β1) Monoclonal antibody specifically detects CD29 (Integrin β1) in Human samples. It is validated for Flow cytometry
Specifications
| CD29 (Integrin β1) | |
| Monoclonal | |
| APC-Cyanine7 | |
| FNRB; GPIIA; ITGB1; MDF2; MSK12; VLA-BETA; VLAB; glycoprotein IIa | |
| Human CTL Cell Line | |
| RUO | |
| Human | |
| Liquid |
| Flow Cytometry | |
| TS2/16 | |
| Aqueous buffered solution containing BSA, protein stabilizer, and ≤0.09% sodium azide. | |
| Mouse | |
| 25 Tests | |
| 3688 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction