Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC Hamster Anti-Mouse CD137 (4-1BB), BD Pharmingen™
Hamster Monoclonal Antibody
Supplier: BD Biosciences 570394
Description
CD137 (4-1BB) Monoclonal antibody specifically detects CD137 (4-1BB) in Mouse samples. It is validated for Flow cytometry
Specifications
CD137 (4-1BB) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Hamster | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
17B5 | |
APC | |
4-1BB; Cd137; ILA; Ly63; Tnfrsf9 | |
Mouse 2PK-3 B Lymphoma Cell Line | |
25 μg | |
21942 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction