Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Mouse anti-Human, Clone: ALi 12-28, Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$280.00

Specifications

Antigen APC
Clone ALi 12-28
Concentration 0.2 mg/ml
Dilution Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Applications Western Blot, Flow Cytometry, ELISA, Immunofluorescence
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB121832
SDP
View Documents
Novus Biologicals
NBP30717920UG
20 μg
Each of 1 for $280.00
Only null left
Add to Cart
 
Description

Description

APC Monoclonal specifically detects APC in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence.
Specifications

Specifications

APC
0.2 mg/ml
Western Blot, Flow Cytometry, ELISA, Immunofluorescence
Unconjugated
Mouse
Cancer, Cell Biology, Cell Cycle and Replication, Metastasis, Tumor Suppressors, Wnt Signaling Pathway
10 mM PBS with 0.05% BSA
324
IgG1 κ
Protein A or G purified
ALi 12-28
Western Blot 1-2 ug/ml, Flow Cytometry 1-2 ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2 ug/ml
Monoclonal
Purified
RUO
Human
adenomatosis polyposis coli, adenomatosis polyposis coli tumor suppressor, adenomatous polyposis coli, adenomatous polyposis coli protein, BTPS2, Deleted in polyposis 2.5, DP2.5DP2, DP3, FPC, GS, Protein APC
Recombinant fragment of human APC protein (around aa 1-433)
Primary
Store at 4°C.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.