Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC Mouse Anti-Human IL-1ra, BD Pharmingen™

Catalog No. BDB570735 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570735 100 Tests
BDB570736 25 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570735 Supplier BD Biosciences Supplier No. 570735
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

IL-1ra Monoclonal antibody specifically detects IL-1ra in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-1ra
Applications Flow Cytometry
Classification Monoclonal
Clone AS17
Conjugate APC
Formulation Aqueous buffered solution containing BSA and ≤0.09% sodium azide.
Gene Alias IL-1RN; IL-1ra; IL1 inhibitor; IL1F3; IL1RN; IRAP; sIL-1ra; icIL-1ra
Host Species Mouse
Immunogen Recombinant Human IL-1 Receptor Antagonist Protein
Purification Method Affinity Chromatography
Quantity 100 Tests
Regulatory Status RUO
Gene ID (Entrez) 3557
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions and unconjugated antibody and free dye were removed.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.