Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APC-PCI Complex Antibody (M36), mFluor Violet 500 SE, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP241380MFV500
Description
APC-PCI Complex Monoclonal antibody specifically detects APC-PCI Complex in Human samples. It is validated for ELISASpecifications
APC-PCI Complex | |
Monoclonal | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Mixture of PCI in complex with PSA (approx. 80%) and PCI cleaved from such a complex (approx. 20%). | |
0.1 mL | |
Primary | |
Store at 4C in the dark. | |
IgG1 κ |
ELISA | |
M36 | |
ELISA | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction