Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC1 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA520941

Catalog No. PIPA520941


Only null left
Add to Cart

Description

Description

A suggested positive control is SK-N-SH cell lysate. PA5-20941 can be used with blocking peptide PEP-1055.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC1, also known as mitotic checkpoint regulator (MCPR), is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

APC1
Polyclonal
Unconjugated
ANAPC1
2610021O03Rik; AI047775; AI853536; ANAPC1; anaphase promoting complex subunit 1; anaphase-promoting complex 1 (meiotic checkpoint regulator); anaphase-promoting complex subunit 1; APC1; AW547281; Cyclosome subunit 1; Mcpr; meiotic check point regulator; mitotic checkpoint regulator; Testis-specific gene 24 protein; TSG24
Rabbit
Antigen affinity chromatography
RUO
17222, 64682
Maintain refrigerated at 2°C to 8°C for up to 3 months. For long term storage store at -20°C
Liquid
Immunohistochemistry, Western Blot
1 mg/mL
PBS with 0.02% sodium azide
P53995, Q9H1A4
ANAPC1
A 17 amino acid peptide near the carboxy terminus of human APC1.
100 μg
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.