Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583639
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583639 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583639 Supplier Invitrogen™ Supplier No. PA583639
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: DLSKGTQHLK SILSKDGVLY VKLRAGQLSY KEDPMGWQSL LAQTVANRNS EARAFKPETI SAFTSDPALL SFAEYFCKPT VNMGQKQEIL DLFSSV Highest antigen sequence indentity to the following orthologs: Mouse - 94%, Rat - 95%.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC1, also known as mitotic checkpoint regulator (MCPR), is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC1
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANAPC1
Gene Accession No. Q9H1A4
Gene Alias 2610021O03Rik; AI047775; AI853536; ANAPC1; anaphase promoting complex subunit 1; anaphase-promoting complex 1 (meiotic checkpoint regulator); anaphase-promoting complex subunit 1; APC1; AW547281; Cyclosome subunit 1; Mcpr; meiotic check point regulator; mitotic checkpoint regulator; Testis-specific gene 24 protein; TSG24
Gene Symbols ANAPC1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ANAPC1. Recombinant protein control fragment (Product #RP-102931).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64682
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.