Learn More
Invitrogen™ APC10 Monoclonal Antibody (8F1D10)

Description
MA5-31623 has been tested in indirect ELISA.
Specifications
Specifications
| Antigen | APC10 |
| Applications | ELISA, Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 8F1D10 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.05% sodium azide |
| Gene | ANAPC10 |
| Gene Accession No. | Q9UM13 |
| Gene Alias | 1500026N15Rik; A830003M23Rik; ANAPC10; anaphase promoting complex subunit 10; anaphase-promoting complex subunit 10; APC10; cyclosome subunit 10; DKFZp564L0562; DOC1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.