Learn More
Invitrogen™ APC10 Polyclonal Antibody

Description
A suggested positive control is mouse heart tissue lysate. PA5-20950 can be used with blocking peptide PEP-1064.
Specifications
Specifications
| Antigen | APC10 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ANAPC10 |
| Gene Accession No. | Q8K2H6, Q9UM13 |
| Gene Alias | 1500026N15Rik; A830003M23Rik; ANAPC10; anaphase promoting complex subunit 10; anaphase-promoting complex subunit 10; APC10; cyclosome subunit 10; DKFZp564L0562; DOC1 |
| Gene Symbols | ANAPC10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.