Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc11 Antibody, Alexa Fluor™ 488, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP176260AF488
Description
Apc11 Polyclonal antibody specifically detects Apc11 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISASpecifications
Apc11 | |
Polyclonal | |
50mM Sodium Borate | |
Rabbit | |
Peptide affinity purified | |
RUO | |
Primary | |
Human, Mouse, Rat | |
Purified |
Western Blot, ELISA | |
Alexa Fluor 488 | |
anaphase promoting complex subunit 11, anaphase promoting complex subunit 11 (yeast APC11 homolog), anaphase-promoting complex subunit 11, APC11 anaphase promoting complex subunit 11 homolog, APC11Hepatocellular carcinoma-associated RING finger protein, Apc11p, Cyclosome subunit 11, HSPC214, MGC882 | |
Antibody was raised against an 18 amino acid synthetic peptide near the center of human APC11. The immunogen is located within amino acids 70 - 120 of APC11. Amino Acid Squence: SLMSREESSRSPDPTPPA | |
0.1 mL | |
Cell Cycle and Replication | |
51529 | |
Store at 4°C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction