Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Apc11 Antibody (OTI3F5) - Azide and BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP271939
Description
Apc11 Monoclonal specifically detects Apc11 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.Specifications
Apc11 | |
Monoclonal | |
LYOPH | |
Western Blot 1:4000, Immunohistochemistry 1:150, Immunocytochemistry/Immunofluorescence 1:100 | |
anaphase promoting complex subunit 11, anaphase promoting complex subunit 11 (yeast APC11 homolog), anaphase-promoting complex subunit 11, APC11 anaphase promoting complex subunit 11 homolog, APC11Hepatocellular carcinoma-associated RING finger protein, Apc11p, Cyclosome subunit 11, HSPC214, MGC882 | |
Mouse | |
20.5 kDa | |
100 μg | |
Primary | |
Reconstitute with PBS (pH 7.3). To use this carrier-free antibody for conjugation experiments, another round of desalting is highly recommended. | |
Store at -20°C. Avoid freeze-thaw cycles. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
OTI3F5 | |
Unconjugated | |
Lyophilized from PBS (pH 7.3) with 8% Trehalose with No Preservative | |
ANAPC11 | |
Full length human recombinant protein of human ANAPC11(NP_001002244) produced in HEk293T cell. | |
Affinity Purified | |
Cell Cycle and Replication | |
51529 | |
Human, Mouse, Rat | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction