Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APC11 Monoclonal Antibody (6F1H6)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA548750
Description
APC11 Monoclonal Antibody for Western Blot, IHC, Flow, ELISA
Together with the cullin protein ANAPC2, constitutes the catalytic component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. The APC/C complex acts by mediating ubiquitination and subsequent degradation of target proteins: it mainly mediates the formation of 'Lys-11'-linked polyubiquitin chains and, to a lower extent, the formation of 'Lys-48'- and 'Lys-63'-linked polyubiquitin chains. May recruit the E2 ubiquitin-conjugating enzymes to the complex. [UniProt].
Specifications
APC11 | |
Monoclonal | |
1 mg/mL | |
PBS with 0.05% sodium azide | |
Q9NYG5 | |
ANAPC11 | |
Purified recombinant fragment of human ANAPC11 (AA: 1-196) expressed in E. Coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
ELISA, Flow Cytometry, Immunohistochemistry, Western Blot | |
6F1H6 | |
Unconjugated | |
ANAPC11 | |
1110011I19Rik; Anapc11; anaphase promoting complex subunit 11; anaphase promoting complex subunit 11 (yeast APC11 homolog); anaphase promoting complex subunit 11 homolog; anaphase-promoting complex subunit 11; APC11; APC11 anaphase promoting complex subunit 11 homolog; Apc11p; Cyclosome subunit 11; hepatocellular carcinoma-associated RING finger protein; HSPC214; R75218; RGD1561880 | |
Mouse | |
Protein G | |
RUO | |
51529 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction