Learn More
Invitrogen™ APC11 Monoclonal Antibody (OTI1F2)

Description
APC11 Monoclonal Antibody for Western Blot, IHC (P)
Specifications
Specifications
| Antigen | APC11 |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Monoclonal |
| Clone | OTI1F2 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | ANAPC11 |
| Gene Accession No. | Q9NYG5 |
| Gene Alias | 1110011I19Rik; Anapc11; anaphase promoting complex subunit 11; anaphase promoting complex subunit 11 (yeast APC11 homolog); anaphase promoting complex subunit 11 homolog; anaphase-promoting complex subunit 11; APC11; APC11 anaphase promoting complex subunit 11 homolog; Apc11p; Cyclosome subunit 11; hepatocellular carcinoma-associated RING finger protein; HSPC214; R75218; RGD1561880 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.