Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC13 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA561154
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA561154 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA561154 Supplier Invitrogen™ Supplier No. PA561154
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MDSEVQRDGR ILDLIDDAWR EDKLPYEDVA IPLNELPEPE QDNGGTTESV KEQEMKWTDL ALQYLHENVP Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC13 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. APC13 promotes the stable association of APC3/Cdc27 and APC6/Cdc16 with the APC/C.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC13
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANAPC13
Gene Accession No. Q9BS18
Gene Alias 1810004D07Rik; ANAPC13; anaphase promoting complex subunit 13; anaphase-promoting complex subunit 13; APC13; cyclosome subunit 13; cylosome subunit 13; SWM1
Gene Symbols ANAPC13
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APC13. Recombinant protein control fragment (Product #RP-96329).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25847
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.