Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA557736
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA557736 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA557736 Supplier Invitrogen™ Supplier No. PA557736
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: PSLFPRVTET LWFNLDRPCV EETELQQQEQ QHQAWLQSIA EKDNNLVPIG KPASEHYDDE EEED Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

ANAPC15 is a component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. In the complex, it plays a role in the release of the mitotic checkpoint complex (MCC) from the APC/C: not required for APC/C activity itself, but promotes the turnover of CDC20 and MCC on the APC/C, thereby participating to the responsiveness of the spindle assembly checkpoint. It is also required for degradation of CDC20.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC15
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Anapc15
Gene Accession No. P60006
Gene Alias 3200002M19Rik; 6330414C15Rik; Anapc15; anaphase promoting complex subunit 15; anaphase prompoting complex C subunit 15; anaphase-promoting complex subunit 15; APC15; C11orf51; C15H11orf51; C21H11orf51; HSPC020; hypothetical protein LOC505356; protein HSPC020 homolog; RGD1311634; uncharacterized protein C11orf51 homolog; Unknown (protein for MGC:153425); zgc:153425
Gene Symbols Anapc15
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APC15. Recombinant protein control fragment (Product #RP-97313).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25906
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.