Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APC15 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA546112
Description
Peptide sequence: EETELQQQEQ QHQAWLQSIA EKDNNLVPIG KPASEHYDDE EEEDDEDDED Sequence homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%; Yeast: 82%; Zebrafish: 90%.
ANAPC15 is a component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. In the complex, it plays a role in the release of the mitotic checkpoint complex (MCC) from the APC/C: not required for APC/C activity itself, but promotes the turnover of CDC20 and MCC on the APC/C, thereby participating to the responsiveness of the spindle assembly checkpoint. It is also required for degradation of CDC20.
Specifications
APC15 | |
Polyclonal | |
Unconjugated | |
Anapc15 | |
3200002M19Rik; 6330414C15Rik; Anapc15; anaphase promoting complex subunit 15; anaphase prompoting complex C subunit 15; anaphase-promoting complex subunit 15; APC15; C11orf51; C15H11orf51; C21H11orf51; HSPC020; hypothetical protein LOC505356; protein HSPC020 homolog; RGD1311634; uncharacterized protein C11orf51 homolog; Unknown (protein for MGC:153425); zgc:153425 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
25906 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
P60006 | |
Anapc15 | |
Synthetic peptide directed towards the N-terminal of human ANAPC15 (aa 26-75). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction