Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ APC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA520944
Description
A suggested positive control is Hela cell lysate. PA5-20944 can be used with blocking peptide PEP-1058.
Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC2 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. Like APC11, APC2 contains cullin and RING finger domains that are thought to be important in regulating ubiquitination activity.
Specifications
| APC2 | |
| Polyclonal | |
| Unconjugated | |
| APC2 | |
| 9230107K09Rik; adenomatosis polyposis coli 2; adenomatous polyposis coli like; adenomatous polyposis coli protein 2; Adenomatous polyposis coli protein-like; AI852447; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; APC2; APC2, WNT signaling pathway regulator; APCL; APC-like; Cyclosome subunit 2; EGK_07151; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406; R75424 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10297, 23805 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| O95996, Q9Z1K7 | |
| APC2 | |
| An 18 amino acid peptide near the center of human APC2. | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction