Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC2 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA516277
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA516277 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA516277 Supplier Thermo Scientific Supplier No. PA516277
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

PA5-16277 targets APC2 in WB applications and shows reactivity with mouse and Human samples. The PA5-16277 immunogen is a peptide from C-terminus of the human APC2.

Cell cycle regulated protein ubiquitination and degradation within subcellular domains is thought to be essential for the normal progression of mitosis. APC2 is a highly conserved component of the anaphase promoting complex/cyclosome (APC/C), a cell cycle-regulated E3 ubiquitin ligase that controls progression through mitosis and the G1 phase of the cell cycle. APC/C is responsible for degrading anaphase inhibitors, mitotic cyclins, and spindle-associated proteins ensuring that events of mitosis take place in proper sequence. The individual APC/C components mRNA and protein levels are expressed at approximately the same levels in most tissues and cell lines, suggesting that they perform their functions as part of a complex. Like APC11, APC2 contains cullin and RING finger domains that are thought to be important in regulating ubiquitination activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC2
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA and 0.09% sodium azide; pH 7.4
Gene APC2
Gene Accession No. O95996, Q9Z1K7
Gene Alias 9230107K09Rik; adenomatosis polyposis coli 2; adenomatous polyposis coli like; adenomatous polyposis coli protein 2; Adenomatous polyposis coli protein-like; AI852447; AL024279; Anapc2; anaphase promoting complex subunit 2; anaphase-promoting complex subunit 2; APC2; APC2, WNT signaling pathway regulator; APCL; APC-like; Cyclosome subunit 2; EGK_07151; Emi4; expressed during mesenchymal induction 4; Imi4; induced during mesenchymal induction 4; KIAA1406; mKIAA1406; R75424
Gene Symbols APC2
Host Species Rabbit
Immunogen A peptide from C-terminus of the human APC2
Purification Method Protein A
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10297, 23805
Target Species Human, Mouse
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.