Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA558373
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA558373 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA558373 Supplier Invitrogen™ Supplier No. PA558373
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: WLYVAMLRMT EDHVLPELNK MTQKDITFVA EFLTEHFNEA PDLYNRKGKY FNVERVGQYL KDEDDDLVSP PNTEGNQWYD FLQN Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.

A large protein complex, termed the anaphase-promoting complex (APC), or the cyclosome, promotes metaphase-anaphase transition by ubiquitinating its specific substrates such as mitotic cyclins and anaphase inhibitor, which are subsequently degraded by the 26S proteasome. Biochemical studies have shown that the vertebrate APC contains eight subunits. The composition of the APC is highly conserved in organisms from yeast to humans. The exact function of this gene product is not known. Two transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ANAPC4
Gene Accession No. Q9UJX5
Gene Alias 2610306D21Rik; ANAPC4; anaphase promoting complex subunit 4; anaphase-promoting complex subunit 4; APC4; cyclosome subunit 4; D5Ertd249e
Gene Symbols ANAPC4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human APC4. Recombinant protein control fragment (Product #RP-96598).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29945
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.