Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APC5 Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501988006
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-8006 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-8006 Supplier Bioss Supplier No. BS7778R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This gene encodes a tetratricopeptide repeat-containing component of the anaphase promoting complex/cyclosome , a large E3 ubiquitin ligase that controls cell cycle progression by targeting a number of cell cycle regulators such as B-type cyclins for 26S proteasome-mediated degradation through ubiquitination. The encoded protein is required for the proper ubiquitination function of APC/C and for the interaction of APC/C with transcription coactivators. It also interacts with polyA binding protein and represses internal ribosome entry site-mediated translation. Multiple transcript variants encoding different isoforms have been found for this gene. These differences cause translation initiation at a downstream AUG and result in a shorter protein , compared to isoform a.

Specifications

Antigen APC5
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene Anapc5
Gene Accession No. Q9UJX4
Gene Alias 2510006G12Rik; AA408751; AA536819; AA986414; Anapc5; anaphase promoting complex subunit 5; Anaphase-promoting complex subunit 5; Anpc5; APC5; Cyclosome subunit 5; MNCb-2778
Gene Symbols Anapc5
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human APC5.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51433
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.