Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA583633
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA583633 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA583633 Supplier Invitrogen™ Supplier No. PA583633
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: KDKLKCYDFD VHTMKTLKNI ISPPWDFREF EVEKQTAEET GLTPLETSRK TPDSRPSLEE TFEIEMNESD MMLETSMSDH ST Highest antigen sequence indentity to the following orthologs: Mouse - 87%, Rat - 89%.

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Cdc16
Gene Accession No. Q13042
Gene Alias 2700071J12Rik; 2810431D22Rik; ANAPC6; Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; Cdc16; CDC16 cell division cycle 16; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16; cell division cycle 16 homolog; cell division cycle protein 16 homolog; CUT9; Cyclosome subunit 6; RP11-569D9.4
Gene Symbols Cdc16
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human CDC16. Recombinant protein control fragment (Product #RP-95122).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8881
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.