Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ APC6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121073
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121073 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121073 Supplier Invitrogen™ Supplier No. PA5121073
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: Jurkat, HepG2, HeLa, A-549, Mouse brain, Mouse ovary, Rat brain. The target is usually found in the following locations: Cytoplasm, centrosome, cytoskeleton, microtubule organizing center, spindle. Immunogen sequence: MLYIGLEYGL TNNSKLAERF FSQALSIAPE DPFVMHEVGV VAFQNGEWKT AEKWFLDALE KIKAIGNEVT VDKWEPLLNN LGHVCRKLKK YAEALDYHRQ ALVLIPQNAS TYSAIGYIHS LMGNFENAVD YFHTALGLRR DDTFSVTMLG HCIEMYIGDS EAYIGADIKD KLKCYDFDVH TMKTLKNIIS PPWDFREFEV EKQTAEETGL TPLETSRKTP DSRPSLEETF EIEMNESDMM LETSMSDHST.

The protein encoded by this gene is structurally very similar to the CDC46 protein from S. cerevisiae, a protein involved in the initiation of DNA replication. The encoded protein is a member of the MCM family of chromatin-binding proteins and can interact with at least two other members of this family. The encoded protein is upregulated in the transition from the G0 to G1/S phase of the cell cycle and may actively participate in cell cycle regulation.
TRUSTED_SUSTAINABILITY

Specifications

Antigen APC6
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.89 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene Cdc16
Gene Accession No. Q13042, Q8R349
Gene Alias 2700071J12Rik; 2810431D22Rik; ANAPC6; Anaphase-promoting complex subunit 6; anaphase-promoting complex, subunit 6; APC6; Cdc16; CDC16 cell division cycle 16; CDC16 cell division cycle 16 homolog; CDC16 homolog; CDC16Hs; cell division cycle 16; cell division cycle 16 homolog; cell division cycle protein 16 homolog; CUT9; Cyclosome subunit 6; RP11-569D9.4
Gene Symbols Cdc16
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 371-620 of human CDC16 (NP_003894.3).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 290875, 69957, 8881
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.