Learn More
Invitrogen™ APC7 Polyclonal Antibody

Description
A suggested positive control is rat kidney tissue lysate. PA5-20948 can be used with blocking peptide PEP-1062.
Specifications
Specifications
| Antigen | APC7 |
| Applications | Immunohistochemistry, Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 0.02% sodium azide |
| Gene | ANAPC7 |
| Gene Accession No. | Q9UJX3, Q9WVM3 |
| Gene Alias | Anapc7; anaphase promoting complex subunit 7; anaphase-promoting complex subunit 7; APC7; AW545589; Cyclosome subunit 7; prediabetic NOD sera-reactive autoantigen; si:ch211-127h20.1 |
| Gene Symbols | ANAPC7 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.