Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
APCDD1 Antibody (2584B), mFluor Violet 610 SE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals FAB10501MFV610
Description
APCDD1 Monoclonal antibody specifically detects APCDD1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
APCDD1 | |
Monoclonal | |
mFluor Violet 610 SE | |
50mM Sodium Borate | |
Rabbit | |
0.1 mL | |
Cancer | |
147495 | |
Store at 4C in the dark. |
Flow Cytometry, CyTOF | |
2584B | |
Flow Cytometry, CyTOF-ready | |
adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1 | |
Mouse myeloma cell line, NS0-derived human APCDD1, Leu27-His492, Accession # Q8J025 | |
RUO | |
Primary | |
Human | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction