Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

APCDD1 Antibody - BSA Free, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody has been used in 1 publication

$192.94 - $517.59

Specifications

Antigen APCDD1
Concentration 1.3 mg/mL
Dilution Western Blot 0.5 ug/ml
Applications Western Blot
Classification Polyclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11092756T
SDP
View Documents
Novus Biologicals
NB11092756SS
0.025 mL
Each for $192.94
Only null left
Add to Cart
 
NB11092756
SDP
View Documents
Novus Biologicals
NB11092756
0.1 mL
Each for $517.59
Only null left
Add to Cart
 
Description

Description

APCDD1 Polyclonal specifically detects APCDD1 in Human, Mouse, Chicken, Reptile samples. It is validated for Western Blot.
Specifications

Specifications

APCDD1
Western Blot 0.5 ug/ml
Polyclonal
Rabbit
Cancer
PBS and 30% Glycerol with 0.1% Sodium Azide
adenomatosis polyposis coli down-regulated 1, Adenomatosis polyposis coli down-regulated 1 protein, B7323, DRAPC1, protein APCDD1
APCDD1
IgG
Affinity Purified
Human, mouse, chicken and turtle.
1.3 mg/mL
Western Blot
Unconjugated
RUO
Human, Mouse, Chicken, Reptile
Q8J025
147495
Synthetic peptide made to an internal portion of human APCDD1 (within residues 400-500). [Swiss-Prot# Q8J025]
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
58 kDa
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.